Learn More
Biomatik Corporation Mouse Thyroxine Binding Globulin (TBG) ELISA Kit
Supplier:  Biomatik Corporation EKU07692
Encompass_Preferred
Mouse Thyroxine Binding Globulin (TBG) ELISA Kit for detecting TBG in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61939. Target Synonyms: SERPINA7; Serpin Peptidase Inhibitor Clade A Member 7; T4-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2938