Learn More
Biomatik Corporation Pig Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07711
Encompass_Preferred
Pig Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit for detecting TIMP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 1.25-80ng/ml. Sensitivity: 0.53ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35624. Gene ID: 396862. Target Synonyms: EPO; CLGI; EPA; HCI; Erythroid Potentiating Activity; Collagenase Inhibitor; Metalloproteinase inhibitor 1; Fibroblast collagenase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2941