Learn More
Biomatik Corporation Mouse Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit
Supplier:  Biomatik Corporation EKU07725
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39876. Gene ID: 21859. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2943