Learn More
Biomatik Corporation Human Toll Like Receptor Adaptor Molecule 1 (TICAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU07763
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 31pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8IUC6. Target Synonyms: TRIF; PRVTIRB; Toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; Proline-rich, vinculin and TIR domain-containing protein B. Restriction: For Research Use Only.
Catalog No. 50-229-2951