Learn More
Biomatik Corporation Pig Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit
Supplier:  Biomatik Corporation EKU07817
Encompass_Preferred
Pig Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15203. Gene ID: 397400. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2957