Learn More
Biomatik Corporation Mouse Tryptophan Hydroxylase 1 (TPH1) ELISA Kit
Supplier:  Biomatik Corporation EKU07928
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: TPH; TPRH; TRPH; Tryptophan 5-Monooxygenase. Restriction: For Research Use Only.
Catalog No. 50-229-2969