Learn More
Biomatik Corporation Rat Vitamin D Binding Protein (DBP) ELISA Kit
Supplier:  Biomatik Corporation EKU08166
Encompass_Preferred
Rat Vitamin D Binding Protein (DBP) ELISA Kit for detecting DBP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.42ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04276. Target Synonyms: GC; VDBG; VDBP; Group-Specific Component; Gc-globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3004