Learn More
Biochain Institute Inc ELISA Kit for Hepatitis B e Antigen, 1 kit/PK
SDP
Supplier:  Biochain Institute Inc KO31006096
Encompass_Preferred
BioChain offers an extensive list of ELISA kits for research use of detection of infectious diseases, such as HBV, HCV, and HIV. The kits are approved and used in clinical diagnosis in other countries, such as China. The assay is a conventional immunological assay where purified antibody is coated on the solid phase of multi-wells. Serum sample, antigen and horseradish peroxidase conjugated antibodies are added to coated wells. The test is special, sensitive, reproducible and easy to operate.
Catalog No. 50-180-836