Learn More
Bioworld ELISA Microplate, U-bottom, clear, medium binding (40/CS), 1 CS
Supplier:  Bioworld 201400031
Encompass_Preferred
ELISA microtitration plate with medium binding capacity96 well flat bottom platesU-bottom, clear, medium bindingManufacturedfrom crystal clear polystyrene, Microlon resinNon-sterileTotal Volume: 323 µlWorking Volume: 40 - 280 µl40/case, 2/sleeve
Catalog No. 50-197-530