Learn More
Actgene Inc ELISA Plate Reader
SDP
Supplier:  Actgene Inc ACTAMPR900B
Encompass_Preferred
ELISA plate reader performs ELISA measurements between the wavelength of 340~900nm with endpoint, two point, and kinetic measurement method.
Catalog No. NC1539335
Only null left
Add to Cart
Request a Quote