Learn More
BioVendor, LLC ELISA SOLUBLE RAGE HUMAN (96 wells)
SDP
Supplier:  BioVendor, LLC RD191116200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, PlasmaSample Requirements50 μl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). TypeSoluble Receptor for Advanced Glycation End Products. Sandwich ELISA, Biotin-labelled antibodyApplicationsSerum, PlasmaSample Requirements50 μl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box).
Catalog No. 50-894-229