Learn More
BioVendor, LLC ELISA S100B HUMAN (96 wells)
SDP
Supplier:  BioVendor, LLC RD192090100R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Cerebrospinal fluidSample Requirements25 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range10–320 pg/mlLimit of DetectionThe measurement range of the assay is 10 – 320 pg/ml (final concentration in a well after dilution).
Catalog No. 50-894-234