Learn More
MTC Bio Sealing film, for ELISA and EIA applications , 10 bags of 10 seals, 100/pk
Supplier:  MTC Bio P1001E
Encompass_Preferred
PureAmp™ Sealing Membranes are compatible with all standard PCR plates. Five types are available for PCR, qPCR, and ELISA applications, pierceable aluminum, standard transparent, optically clear sticky adhesive, optically clear pressure-sensitive adhesive, and low-adhesive EZ-peel all of which are guaranteed to keep evaporation rates below 5%. RNase, DNase, DNA, and PCR inhibitor free.
Catalog No. 50-226-8477