Learn More
BioVendor, LLC ELISA SHLA-G (96 wells)
SDP
Supplier:  BioVendor, LLC RD194070100R
Encompass
TypeSandwich ELISA, HRP-labelled antibodyApplicationsPlasma-EDTA, Amniotic fluid, Cell culture supernatantSample Requirements25 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range3.91–125 Units/mlLimit of Detection0.6 Units/ml
Catalog No. 50-894-235