Learn More
Cell Signaling Technology ELISA Wash Buffer, 20X Solution, 125 mL
SDP
Catalog No. 502050446
Encompass
ELISA Wash Buffer (20X) is specifically formulated for use with both FastScan™ and PathScan™ ELISA Kits. It serves as the recommended buffer for all wash steps within the protocols for these kits.
  • Supplied as a 20X solution
  • Produces up to 2.5 L of 1X wash buffer
  • Compatible with specified ELISA kit protocols
  • Requires dilution with double distilled H2O
Catalog No. 50-205-0446 Supplier Cell Signaling Technology Supplier No. 9801P2
ELISA Wash Buffer (20X) is specifically formulated for use with both FastScan™ and PathScan™ ELISA Kits. It serves as the recommended buffer for all wash steps within the protocols for these kits.
  • Supplied as a 20X solution
  • Produces up to 2.5 L of 1X wash buffer
  • Compatible with specified ELISA kit protocols
  • Requires dilution with double distilled H2O