Learn More
Mabtech Inc Elispot Pvdf Plates, Clear, 10 Plates
Catalog No. NC1856932
Encompass
These MSIP plates are sterile MultiscreenHTSIP plates suitable for ELISpot assays. Plates are clear and equipped with an underdrain. For optimal binding of capture antibody, it is recommended to pre-wet, i.e. activate, the PVDF membrane with ethanol.
Catalog No. NC1856932 Supplier Mabtech Inc Supplier No. 3654TP10
Add to Cart
Add to Cart
These MSIP plates are sterile MultiscreenHTSIP plates suitable for ELISpot assays. Plates are clear and equipped with an underdrain. For optimal binding of capture antibody, it is recommended to pre-wet, i.e. activate, the PVDF membrane with ethanol.