Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD ELISPOT Sets

Catalog No. BDB551018 Shop All BD Biosciences Products
Change view
Click to view available options
Antigen:
Granzyme B
IFN-gamma
IL-10
IL-12 p70
IL-2
IL-4
IL-5
IL-6
TNF
Cross Reactivity:
Human
Mouse
Product Type:
Human Granzyme B ELISPOT set
Human IFN-gamma ELISPOT set
Human IL-10 ELISPOT set
Human IL-12 p70 ELISPOT set
Human IL-2 ELISPOT set
Human IL-4 ELISPOT set
Human IL-5 ELISPOT set
Mouse IFN-gamma ELISPOT set
Mouse IL-2 ELISPOT set
Mouse IL-4 ELISPOT set
Mouse IL-5 ELISPOT set
Mouse IL-6 ELISPOT set
Mouse TNF ELISPOT set
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Antigen Cross Reactivity Product Type
BDB551018 IL-10 Human Human IL-10 ELISPOT set
BDB551849 IFN-gamma Human Human IFN-gamma ELISPOT set
BDB551075 IL-5 Mouse Mouse IL-5 ELISPOT set
BDB551085 IL-5 Human Human IL-5 ELISPOT set
BDB551491 TNF Mouse Mouse TNF ELISPOT set
BDB551084 IL-4 Human Human IL-4 ELISPOT set
BDB552572 Granzyme B Human Human Granzyme B ELISPOT set
BDB551076 IL-2 Mouse Mouse IL-2 ELISPOT set
BDB551017 IL-4 Mouse Mouse IL-4 ELISPOT set
BDB551282 IL-2 Human Human IL-2 ELISPOT set
BDB551083 IFN-gamma Mouse Mouse IFN-gamma ELISPOT set
BDB552574 IL-12 p70 Human Human IL-12 p70 ELISPOT set
BDB552567 IL-6 Mouse Mouse IL-6 ELISPOT set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. BDB551018 Supplier BD Supplier No. 551018
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations

Sufficient capture antibody, detection antibody, and streptavidin-horseradish peroxidase for 10 x 96 wells

  • High sensitivity
  • High throughput, high content analysis
  • Minimal volume of biological material required
  • Suitable for frozen/thawed biological samples
  • Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis

TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Cross Reactivity Human
Includes Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates
Quantity 10 x 96 Tests
Content And Storage 4°C
For Use With (Application) Cloning, proliferation assays, flow cytometry or other methods of analysis
Product Type Human IL-10 ELISPOT set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.