Learn More
Fisherbrand™ Elite™ Adjustable-Volume Pipetters
SCI_ED
Catalog No. S98635A Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
100 to 1000 μL
2 to 20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S98635A 100 to 1000 μL
S98632A 2 to 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S98635A
This item is not returnable. View return policy
Includes
Calibration certificate and in-lab calibration tool.
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
  • Fully autoclavable — entire pipetter can be steam autoclaved
  • Extremely low plunger forces — work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
  • Optimal fit with SureOne™ pipetter tips — best pipetting performance
  • Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Warranty and Services
Three years.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
For Use With SureOne™ Pipette Tip
Product Line Elite
Type Micro Pipette
Volume (Metric) 100 to 1000 μL