Learn More
Perkin Elmer US LLC Elite Biodiesel TG Capillary Column, 14 m x 0.53 mm I.D., 0.16 µm
Catalog No. 502117332
Encompass_Preferred
Note that the column has a total length of 16 m and is made of specially treated stainless steel. Temperature limits -60 to 380/430°C.
Catalog No. 50-211-7332 Supplier Perkin Elmer US LLC Supplier No. N9306071
Note that the column has a total length of 16 m and is made of specially treated stainless steel. Temperature limits -60 to 380/430°C.