Learn More
StatLab Elite PAP Pen
SDP
Catalog No. 21010250
Change view
Click to view available options
Type:
Hydrophobic Pen
Catalog No. Type
21-010-250 Hydrophobic Pen
1 options
This item is not returnable. View return policy
Catalog No. 21-010-250 Supplier StatLab Supplier No. ACR012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hydrophobic Liquid Blocker.
  • Broad point
  • Suitable for both immunohistochemical and fluorescent staining
  • Hydrophobic properties allow the user to draw barriers on the slide to confine the flow of reagents to a defined area
Specifications
Type Hydrophobic Pen