Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Elite Plus Silver Immunochemical Rapid Fecal Occult Blood Test
SDP

Catalog No. 21520137
Change view
Click to view available options
Quantity:
30 Tests/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-137 30 Tests/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-520-137 Supplier Cenmed™ Supplier No. 4263001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Elite Plus iFOB is designed to detect lower levels of fecal occult blood than standard guaiac tests

  • Ready to use, provides results within minutes
  • Packaged in a bulk pack for conveneince and use within a hospital or doctors office
  • Easy to read and interpret
  • Materials included are: 30 Test Devices, 30 Sample Collection Tubes, 1 Package Insert, 1 Bottle of FOB Buffer
  • Results in 5 to 10 minutes; Positive results may become visible sooner
  • Recommended for hospitals performing iFOBT screenings
  • Does not include mailers
  • Cutoff is 50 ng hHb/mL

Specifications

Accuracy >98%
Content And Storage Room Temperature
Format Bulk Pack
Sensitivity 50 ng hHB/mL
For Use With (Application) Immunological fecal occult Blood test
Material 30 Fecal Occult Blood Test Cassettes, 30 Collection Tube Buffers, 1 Insert
Quantity 30 Tests/Box
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.