Learn More
Perkin Elmer US LLC Elite-1 Capillary Column - 30 m x 0.53 mm I.D. x 1.50 μm
Catalog No. 502105474
Encompass_Preferred
For use at temperatures ranging from -60°C to 310°C/330°C.
Catalog No. 50-210-5474 Supplier Perkin Elmer US LLC Supplier No. N9316050
Add to Cart
Add to Cart
Request a Quote
For use at temperatures ranging from -60°C to 310°C/330°C.