Learn More
Perkin Elmer US LLC Elite-1 Capillary Column - 60 m x 0.32 mm I.D. x 5.00 μm
Catalog No. 502105448
Encompass_Preferred
For use at temperatures ranging from -60°C to 260°C/280°C.
Catalog No. 50-210-5448 Supplier Perkin Elmer US LLC Supplier No. N9316031
For use at temperatures ranging from -60°C to 260°C/280°C.