Learn More
Perkin Elmer US LLC Elite-1 Metal Capillary Column, 15 m, 0.25 mm I.D., 0.25 µm df
Catalog No. 502112501
Encompass_Preferred
Elite-1 100% dimethyl polysiloxane, columns are highly versatile, ideal for the analysis of a wide range of non-polar compounds including petrochemical samples, solvents, essential oils and chemicals. Equivalent to USP G1, G2, G38 phases Temperature Limits: -60 to 360/430°C Column is coiled to a 4.5 inch diameter
Catalog No. 50-211-2501 Supplier Perkin Elmer US LLC Supplier No. N9303454
Elite-1 100% dimethyl polysiloxane, columns are highly versatile, ideal for the analysis of a wide range of non-polar compounds including petrochemical samples, solvents, essential oils and chemicals. Equivalent to USP G1, G2, G38 phases Temperature Limits: -60 to 360/430°C Column is coiled to a 4.5 inch diameter