Learn More
Perkin Elmer US LLC Elite-1ms Capillary Column - 30 m x 0.32 mm I.D. x 0.25 µm
Catalog No. 502117156
Encompass_Preferred
For use at temperatures ranging from -60°C to 330°C/350°C.
Catalog No. 50-211-7156 Supplier Perkin Elmer US LLC Supplier No. N9305647
For use at temperatures ranging from -60°C to 330°C/350°C.