Learn More
Perkin Elmer US LLC Elite-1ms Capillary Column - 60 m x 0.32 mm I.D. x 1.00 µm
Catalog No. 502117152
Encompass_Preferred
For use at temperatures ranging from -60°C to 330°C/350°C.
Catalog No. 50-211-7152 Supplier Perkin Elmer US LLC Supplier No. N9305653
For use at temperatures ranging from -60°C to 330°C/350°C.