Learn More
Perkin Elmer US LLC Elite-200 Capillary Column - 30 m x 0.32 mm I.D. x 1.00 µm
Catalog No. 502104566
Encompass_Preferred
For use at temperatures ranging from -20°C to 290°C/310°C.
Catalog No. 50-210-4566 Supplier Perkin Elmer US LLC Supplier No. N9316630
For use at temperatures ranging from -20°C to 290°C/310°C.