Learn More
Perkin Elmer US LLC Elite-5 Capillary Column - 30 m x 0.53 mm I.D. x 1.50 μm
Catalog No. 502105511
Encompass_Preferred
For use at temperatures ranging from -60°C to 310°C/330°C.
Catalog No. 50-210-5511 Supplier Perkin Elmer US LLC Supplier No. N9316100
For use at temperatures ranging from -60°C to 310°C/330°C.