Learn More
Perkin Elmer US LLC Elite-5 Capillary Column - 60 m x 0.32 mm I.D. x 1.00 μm
Catalog No. 502105501
Encompass_Preferred
For use at temperatures ranging from -60°C to 320°C/340°C.
Catalog No. 50-210-5501 Supplier Perkin Elmer US LLC Supplier No. N9316091
For use at temperatures ranging from -60°C to 320°C/340°C.