Learn More
Perkin Elmer US LLC Elite-5 Capillary Column - 60 m x 0.25 mm I.D. x 0.25 μm
Catalog No. 502105486
Encompass_Preferred
For use at temperatures ranging from -60°C to 330°C/350°C.
Catalog No. 50-210-5486 Supplier Perkin Elmer US LLC Supplier No. N9316079
For use at temperatures ranging from -60°C to 330°C/350°C.