Learn More
Perkin Elmer US LLC Elite-5ht Capillary Column - 15 m x 0.32 mm I.D. x 0.10 µm
Catalog No. 502113010
Encompass_Preferred
Elite-5ht Capillary Column - 15 m x 0.32 mm I.D. x 0.10 µm
Catalog No. 50-211-3010 Supplier Perkin Elmer US LLC Supplier No. N9316274
Elite-5ht Capillary Column - 15 m x 0.32 mm I.D. x 0.10 µm