Learn More
Perkin Elmer US LLC Elite-5ms Capillary Column - 20 m x 0.18 mm I.D. x 0.18 µm
Catalog No. 502111410
Encompass_Preferred
For use at temperatures ranging from -60°C to 325°C/340°C.
Catalog No. 50-211-1410 Supplier Perkin Elmer US LLC Supplier No. N9316276
For use at temperatures ranging from -60°C to 325°C/340°C.