Learn More
Perkin Elmer US LLC Elite-5ms Capillary Column - 30 m x 0.32 mm I.D. x 1.00 µm
Catalog No. 502111420
Encompass_Preferred
For use at temperatures ranging from -60°C to 325°C/350°C.
Catalog No. 50-211-1420 Supplier Perkin Elmer US LLC Supplier No. N9316294
For use at temperatures ranging from -60°C to 325°C/350°C.