Learn More
Perkin Elmer US LLC Elite-5ms Capillary Column - 50 m x 0.20 mm I.D. x 0.33 µm
Catalog No. 502111423
Encompass_Preferred
For use at temperatures ranging from -60°C to 330°C/350°C.
Catalog No. 50-211-1423 Supplier Perkin Elmer US LLC Supplier No. N9316303
For use at temperatures ranging from -60°C to 330°C/350°C.