Learn More
Perkin Elmer US LLC Elite-624ms Capillary Column, 30m x 0.32mm, 1.80µm
Catalog No. 502112948
Encompass_Preferred
Temperature range: -20 to 320 degrees celsius. Similar to USP G43 phase.
Catalog No. 50-211-2948 Supplier Perkin Elmer US LLC Supplier No. N9315069
Temperature range: -20 to 320 degrees celsius. Similar to USP G43 phase.