Learn More
Perkin Elmer US LLC Elite-624ms Capillary Column, 30m x 0.25mm, 1.40µm
Catalog No. 502112950
Encompass_Preferred
Temperature range: -20 to 320 degrees celsius. Similar to USP G43 phase.
Catalog No. 50-211-2950 Supplier Perkin Elmer US LLC Supplier No. N9315068
Temperature range: -20 to 320 degrees celsius. Similar to USP G43 phase.