Learn More
Perkin Elmer US LLC Elite-Alumina PLOT Capillary Column - 30 m x 0.53 mm I.D. x 10.00 µm
Catalog No. 502111424
Encompass_Preferred
For the analysis of light hydrocarbons at temperatures ranging from -60°C to 200°C.
Catalog No. 50-211-1424 Supplier Perkin Elmer US LLC Supplier No. N9316304
Add to Cart
Add to Cart
Request a Quote
For the analysis of light hydrocarbons at temperatures ranging from -60°C to 200°C.