Learn More
Perkin Elmer US LLC Elite-FFAP Capillary Column - 15 m x 0.53 mm I.D. x 1.00 µm
Catalog No. 502111443
Encompass_Preferred
For the analysis of free fatty acids at temperatures ranging from 40°C to 240°C/250°C.
Catalog No. 50-211-1443 Supplier Perkin Elmer US LLC Supplier No. N9316357
For the analysis of free fatty acids at temperatures ranging from 40°C to 240°C/250°C.