Learn More
Perkin Elmer US LLC Elite-FFAP Capillary Column - 30 m x 0.32 mm I.D. x 0.25 µm
Catalog No. 502111446
Encompass_Preferred
For the analysis of free fatty acids at temperatures ranging from 40°C to 250°C.
Catalog No. 50-211-1446 Supplier Perkin Elmer US LLC Supplier No. N9316354
For the analysis of free fatty acids at temperatures ranging from 40°C to 250°C.