Learn More
Perkin Elmer US LLC Elite-PONA Capillary Column - 100 m x 0.25 mm I.D. x 0.50 μm
Catalog No. 502105374
Encompass_Preferred
For the detailed analysis of petroleum naphtha at temperatures ranging from -60°C to 300°C/320°C.
Catalog No. 50-210-5374 Supplier Perkin Elmer US LLC Supplier No. N9316015
For the detailed analysis of petroleum naphtha at temperatures ranging from -60°C to 300°C/320°C.