Learn More
Perkin Elmer US LLC Elite-Q PLOT Capillary Column - 30 m x 0.32 mm I.D. x 10.00 µm
Catalog No. NC1901327
Encompass_Preferred
For the analysis of light gases and hydrocarbons at temperatures ranging from -60°C to 250°C.
Catalog No. NC1901327 Supplier Perkin Elmer US LLC Supplier No. N9316359
For the analysis of light gases and hydrocarbons at temperatures ranging from -60°C to 250°C.