Learn More
MilliporeSigma™ Electrode Deionized Modules
Catalog No. zlx0edi03 Shop All MilliporeSigma Products
Change view
Click to view available options
:
Electrode Deionized Module
Catalog No.
ZLX0EDI03 Electrode Deionized Module
1 options
Catalog No. ZLX0EDI03 Supplier MilliporeSigma™ Supplier No. ZLX0EDI03
Only null left
Add to Cart
Add to Cart
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories -- in both validated and non-validated environments.
The Elix™ EDI module has been designed to continuously removes remaining ions, by electrodeionization, for consistently superior quality pure water, without any maintenance and at 3 L/h production flow rate.
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Deionized Module