Learn More
Braintree Scientific Elizabethan Collar for Rats up to 450g
SDP
Catalog No. NC9263311
Encompass
Elizabethan Collars. Designed to prevent animals from grooming and to protect surgical or treatment sites. All collars are manufactured from a low-density polyethylene, for smaller animals a lightweight material is used. Collars close with Velcro fasteners providing adjustable and secure fit for individual animals. Neck openings are bound with cotton jersey for comfort and light padding. **Sold for research purposes only and not for human use**
Catalog No. NC9263311 Supplier Braintree Scientific Supplier No. ECR1
Elizabethan Collars. Designed to prevent animals from grooming and to protect surgical or treatment sites. All collars are manufactured from a low-density polyethylene, for smaller animals a lightweight material is used. Collars close with Velcro fasteners providing adjustable and secure fit for individual animals. Neck openings are bound with cotton jersey for comfort and light padding. **Sold for research purposes only and not for human use**