Learn More
ATCC Elizabethkingia meningoseptica
Supplier:  ATCC 13253
Encompass
Type strain. Formerly Chryseobacterium meningosepticum. Group A. Quality control strain for BBL products and VITEK 2 GN Comprehensive QC Set.
Catalog No. 50-238-4000