Learn More
Elkay Plastics Company Bag Reclosable Clear with Zip Closure 2 mil 5 X 8 1,000 per Case
Catalog No. NC0954876
Encompass_Preferred
LK Clear Line Zipper Reclosable bags provide excellent protection, convenience and economy. The zipper type seal closes securely and is easy to open and close time after time. These bags are excellent for storing, displaying, protecting and shipping all types of product. Because they are made only with virgin resin, they meet FDA requirements for contact with food products. Bags under 15" wide are packed in cartons of 1,000 with inner packs of 100 bags each for added convenience and to help reduce waste.
Catalog No. NC0954876 Supplier Elkay Plastics Company Supplier No. F20508
LK Clear Line Zipper Reclosable bags provide excellent protection, convenience and economy. The zipper type seal closes securely and is easy to open and close time after time. These bags are excellent for storing, displaying, protecting and shipping all types of product. Because they are made only with virgin resin, they meet FDA requirements for contact with food products. Bags under 15" wide are packed in cartons of 1,000 with inner packs of 100 bags each for added convenience and to help reduce waste.