Learn More
Aviva Systems Biology Corp ELK1P1 ANTIBODY 50UG/50UL
SDP
Supplier:  Aviva Systems Biology Corp ARP51071_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC647102. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human LOC647102. Alternate names: ELK1P1ApplicationWB. Species reactivity: Human
Catalog No. 50-285-553