Learn More
Aviva Systems Biology Corp Elk3 Antibody - N-terminal region (ARP38590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Elk3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: D430049E23Rik, Erp, Etrp, Net, Sap-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-747