Learn More
Aviva Systems Biology Corp ELK3 Antibody - middle region (P100856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ELK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ELK3. Alternate names: ERP, NET, SAP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62745