Learn More
Aviva Systems Biology Corp ELL Antibody - N-terminal region (ARP38813_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38813_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ELL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ELL. Alternate names: MEN, ELL1, PPP1R68, C19orf17ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-56812