Learn More
Medchemexpress LLC Ellagic acid | 476-66-4 | 99.9% | 5 G
SDP
Catalog No. 5000405680
Encompass_Preferred
Ellagic acid is a natural antioxidant that acts as a potent and ATP-competitive inhibitor of CK2 and SHP2. It has an IC50 of 40 nM and a Ki of 20 nM.
  • Natural antioxidant
  • Potent ATP-competitive inhibitor of CK2
  • Potent ATP-competitive inhibitor of SHP2
  • IC50 of 40 nM
  • Ki of 20 nM
Catalog No. 50-004-05680 Supplier Medchemexpress LLC Supplier No. HYB01835G
Ellagic acid is a natural antioxidant that acts as a potent and ATP-competitive inhibitor of CK2 and SHP2. It has an IC50 of 40 nM and a Ki of 20 nM.
  • Natural antioxidant
  • Potent ATP-competitive inhibitor of CK2
  • Potent ATP-competitive inhibitor of SHP2
  • IC50 of 40 nM
  • Ki of 20 nM